This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01060460_m1
See other AUH GE Assays ›
Gene Symbol:
AUH
Gene Name:
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases:
-
Chromosome Location:
Chr.9: 91213815 - 91361969 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01060460_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AUH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306190.1 NP_001293119.1 4 - 5 590 72
NM_001698.2 NP_001689.1 5 - 6 632 72
XM_005252066.3 XP_005252123.1 6 - 7 675 72
XM_005252067.4 XP_005252124.1 6 - 7 677 72
XM_005252069.4 XP_005252126.1 6 - 7 678 72
XM_005252072.2 XP_005252129.1 5 - 6 632 72
XM_006717150.3 XP_006717213.1 5 - 6 591 72
XM_011518800.2 XP_011517102.1 6 - 7 679 72
XM_011518802.2 XP_011517104.1 6 - 7 701 72
XM_017014849.1 XP_016870338.1 5 - 6 632 72
XM_017014850.1 XP_016870339.1 6 - 7 772 72
XM_017014851.1 XP_016870340.1 6 - 7 701 72
GenBank mRNA AK302453.1 - 5 - 6 603 72
BC020722.1 - 4 - 5 534 4656960 72
BG706374.1 - 5 - 6 628 72
X79888.1 - 5 - 6 601 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010762.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001324.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AUH

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

leucine catabolic process
fatty acid beta-oxidation
branched-chain amino acid catabolic process
mRNA 3'-UTR binding
enoyl-CoA hydratase activity
methylglutaconyl-CoA hydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000475023.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000475023.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000475023.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009767.hg.1 ENST00000408239.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010349.hg.1 ENST00000408341.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016475.hg.1 TC09001324.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09016475.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016475.hg.1 ENST00000375731.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016475.hg.1 ENST00000422391.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016475.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0300010349.hg.1 ENST00000408341.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay PSR0900173189.hg.1 TC0900010762.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800009767.hg.1 ENST00000408239.3 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000303617.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000375731.8 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000473695.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000475023.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000475023.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000475023.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 ENST00000478465.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001306190.1 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq
Affymetrix Human Clariom D Assay TC0900010762.hg.1 NM_001698.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0