This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061512_m1
See other CHD1 GE Assays ›
Gene Symbol:
CHD1
Gene Name:
chromodomain helicase DNA binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 98855204 - 98929014 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061512_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270.2 NP_001261.2 34 - 35 4929 142
XM_005271866.4 XP_005271923.1 36 - 37 5669 142
XM_005271867.4 XP_005271924.1 35 - 36 5405 142
XM_017008991.1 XP_016864480.1 20 - 21 2729 142
XM_017008992.1 XP_016864481.1 20 - 21 2574 142
GenBank mRNA AF006513.1 - 34 - 35 4944 142
AK094871.1 - 3 - 4 969 142
BC117134.1 - 34 - 35 4919 40125685 142
BQ436172.1 - 6 - 7 693 142
BX648767.1 - 9 - 10 1447 142

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD1

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
DNA duplex unwinding
positive regulation by host of viral transcription
DNA binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
methylated histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline