This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061653_g1
See other N6AMT1 GE Assays ›
Gene Symbol:
N6AMT1
Gene Name:
N-6 adenine-specific DNA methyltransferase 1 (putative)
Gene Aliases:
C21orf127, HEMK2, MTQ2, N6AMT, PRED28, m.HsaHemK2P
Chromosome Location:
Chr.21: 28571095 - 28885373 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061653_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N6AMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013240.5 NP_037372.4 5 - 6 564 138
NM_182749.4 NP_877426.4 4 - 5 480 138
NR_047510.2 5 - 6 564 138
GenBank mRNA AF139682.1 - 5 - 6 565 138
AK225515.1 - 5 - 6 558 138
AK314100.1 - 5 - 6 556 138
BC011554.1 - 4 - 5 461 4640080 138

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007846.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000350.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene N6AMT1

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

translational termination
protein methylation
positive regulation of cell growth
methylation
nucleic acid binding
protein binding
protein methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21003963.hg.1 ENST00000303775.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21003963.hg.1 ENST00000351429.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21003963.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21003963.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21003963.hg.1 TC21000350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 TC2100007846.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay PSR2100152885.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 TC2100007846.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100152880.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0