This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061790_m1
See other SUMF1 GE Assays ›
Gene Symbol:
SUMF1
Gene Name:
sulfatase modifying factor 1
Gene Aliases:
AAPA3037, FGE, UNQ3037
Chromosome Location:
Chr.3: 4034714 - 4467282 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061790_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUMF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164674.1 NP_001158146.1 4 - 5 684 82
NM_001164675.1 NP_001158147.1 5 - 6 759 82
NM_182760.3 NP_877437.2 5 - 6 759 82
XM_011533624.2 XP_011531926.1 5 - 6 750 82
XM_011533625.2 XP_011531927.1 5 - 6 750 82
XM_011533626.2 XP_011531928.1 5 - 6 750 82
XM_017006252.1 XP_016861741.1 5 - 6 750 82
XM_017006253.1 XP_016861742.1 4 - 5 672 82
XM_017006254.1 XP_016861743.1 5 - 6 750 82
XM_017006255.1 XP_016861744.1 5 - 6 750 82
GenBank mRNA AB448737.1 - 5 - 6 730 82
AK057983.1 - 6 - 7 658 82
AK075459.1 - 5 - 6 741 82
AK302018.1 - 4 - 5 652 82
AY208752.1 - 5 - 6 722 82
AY323910.1 - 5 - 6 722 82
AY358092.1 - 5 - 6 739 82
BC110862.1 - 5 - 6 730 5760490 82
BC121122.2 - 5 - 6 751 40120670 82
BC121123.2 - 5 - 6 751 40120671 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013926.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001125.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SUMF1

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
post-translational protein modification
oxidation-reduction process
oxidoreductase activity
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000431226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0