This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061792_m1
See other SUMF1 GE Assays ›
Gene Symbol:
SUMF1
Gene Name:
sulfatase modifying factor 1
Gene Aliases:
AAPA3037, FGE, UNQ3037
Chromosome Location:
Chr.3: 4034714 - 4467282 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061792_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUMF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164674.1 NP_001158146.1 7 - 8 971 68
NM_182760.3 NP_877437.2 8 - 9 1046 68
GenBank mRNA AB448737.1 - 8 - 9 1017 68
AK057983.1 - 9 - 10 945 68
AK075459.1 - 8 - 9 1028 68
AK295240.1 - 4 - 5 637 68
AK302018.1 - 7 - 8 939 68
AY208752.1 - 8 - 9 1009 68
AY323910.1 - 8 - 9 1009 68
BC017005.2 - 3 - 4 285 68
BC110862.1 - 8 - 9 1017 5760490 68
BC121122.2 - 8 - 9 1038 40120670 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013926.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001125.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SUMF1

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
post-translational protein modification
oxidation-reduction process
oxidoreductase activity
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000431226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0