This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061979_g1
See other CNTFR GE Assays ›
Gene Symbol:
CNTFR
Gene Name:
ciliary neurotrophic factor receptor
Gene Aliases:
-
Chromosome Location:
Chr.9: 34551432 - 34590386 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061979_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNTFR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001207011.1 NP_001193940.1 5 - 6 608 76
NM_001842.4 NP_001833.1 4 - 5 621 76
NM_147164.2 NP_671693.1 5 - 6 732 76
XM_017014260.1 XP_016869749.1 6 - 7 948 76
XM_017014261.1 XP_016869750.1 6 - 7 921 76
XM_017014262.1 XP_016869751.1 6 - 7 865 76
XM_017014263.1 XP_016869752.1 5 - 6 706 76
XM_017014264.1 XP_016869753.1 5 - 6 1250 76
XM_017014265.1 XP_016869754.1 4 - 5 566 76
GenBank mRNA AK127444.1 - 1 - 2 1713 76
AL537375.3 - 5 - 6 607 76
BC001492.2 - 4 - 5 578 3510004 76
BT019824.1 - 3 - 4 438 76
M73238.1 - 5 - 6 701 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009905.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001038.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK127444.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CNTFR

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
brainstem development
signal transduction
nervous system development
sex differentiation
positive regulation of cell proliferation
negative regulation of neuron apoptotic process
skeletal muscle organ development
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
receptor binding
protein binding
cytokine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09013602.hg.1 TC09001038.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09013602.hg.1 ENST00000351266.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09013602.hg.1 ENST00000378980.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09013602.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09013602.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09013602.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay TC0900009905.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000351266.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000378980.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000417345.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 ENST00000610543.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 NM_001207011.1 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 NM_001842.4 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 NM_147164.2 Refseq
Affymetrix Human Clariom D Assay PSR0900167874.hg.1 TC0900009905.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-2vf2v:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline