This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01062473_g1
See other GFI1B GE Assays ›
Gene Symbol:
GFI1B
Gene Name:
growth factor independent 1B transcriptional repressor
Gene Aliases:
BDPLT17, ZNF163B
Chromosome Location:
Chr.9: 132945545 - 132991697 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01062473_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GFI1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004188.6 NP_004179.3 9 - 10 1475 88
XM_011519070.2 XP_011517372.1 7 - 8 1366 88
GenBank mRNA AY428733.1 - 5 - 6 801 88
BC043371.1 - 9 - 10 1475 5170920 88
BC048254.1 - 4 - 5 681 88
CR536546.1 - 4 - 5 650 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009043.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GFI1B

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell proliferation
covalent chromatin modification
hemopoiesis
regulation of histone H3-K4 methylation
RNA polymerase II transcription factor binding
DNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000339463.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000372123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000631293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 ENST00000631293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_001135031.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009043.hg.1 NM_004188.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0