This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01062599_m1
See other YIF1B GE Assays ›
Gene Symbol:
YIF1B
Gene Name:
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases:
FinGER8
Chromosome Location:
Chr.19: 38303560 - 38315976 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01062599_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YIF1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039673.2 NP_001034762.1 1 - 2 98 114
NM_001145463.1 NP_001138935.1 1 - 2 98 114
XM_017027449.1 XP_016882938.1 1 - 2 108 114
GenBank mRNA AW630507.1 - 1 - 2 98 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010597.hg
This assay targets the same location as HTA transcript cluster(s) TC19001497.hg

Additional Information:

This assay is in the 5'UTR region of the NM_001039673.2,NM_001145463.1,XM_017027449.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19020324.hg.1 ENST00000337679.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020324.hg.1 ENST00000339413.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020324.hg.1 NM_001039672.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020324.hg.1 NM_001039673.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020324.hg.1 NM_001145463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020324.hg.1 TC19001497.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19020326.hg.1 ENST00000337679.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020326.hg.1 ENST00000339413.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020326.hg.1 NM_001039672.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020326.hg.1 NM_001039673.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020326.hg.1 NM_001145463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020326.hg.1 TC19001497.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 ENST00000329420.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 ENST00000337679.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 ENST00000339413.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 ENST00000392124.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 NM_001039671.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 NM_001039672.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 NM_001039673.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 NM_001145461.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 NM_001145462.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 NM_001145463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19020318.hg.1 TC19001497.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000337679.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000339413.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000589644.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000591755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000592694.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 NM_001039672.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 NM_001039673.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 NM_001145463.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 TC1900010597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000337679.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000339413.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000589644.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000591755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 ENST00000592694.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 NM_001039672.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 NM_001039673.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187161.hg.1 NM_001145463.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000329420.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000337679.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000339413.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000392124.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000585563.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000587039.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000587361.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000588002.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000589247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000589644.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000591755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000591784.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000592246.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000592694.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001039671.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001039672.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001039673.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001145461.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001145462.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001145463.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 TC1900010597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000329420.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000337679.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000339413.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000392124.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000585563.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000587039.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000587361.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000588002.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000589247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000589644.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000591755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000591784.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000592246.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 ENST00000592694.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001039671.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001039672.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001039673.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001145461.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001145462.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187148.hg.1 NM_001145463.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000329420.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000337679.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000339413.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000392124.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000585563.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000587039.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000587361.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000588002.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000589247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000589644.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000591755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000591784.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000592246.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000592694.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001039671.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001039672.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001039673.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001145461.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001145462.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001145463.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 TC1900010597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000329420.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000337679.12 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000339413.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000392124.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000585563.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000587039.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000587361.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000588002.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000589247.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000589644.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000591755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000591784.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000592246.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 ENST00000592694.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001039671.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001039672.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001039673.2 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001145461.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001145462.1 Refseq
Affymetrix Human Clariom D Assay PSR1900187147.hg.1 NM_001145463.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0