This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01064347_m1
See other TRNT1 GE Assays ›
Gene Symbol:
TRNT1
Gene Name:
tRNA nucleotidyl transferase 1
Gene Aliases:
CCA1, CGI-47, MtCCA, RPEM, SIFD
Chromosome Location:
Chr.3: 3126916 - 3153435 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01064347_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRNT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001302946.1 NP_001289875.1 1 - 2 78 54
NM_182916.2 NP_886552.2 1 - 2 78 54
XM_011533778.2 XP_011532080.1 1 - 2 78 54
GenBank mRNA AK290411.1 - 1 - 2 43 54
AL834397.1 - 1 - 2 42 54
BC005184.1 - 1 - 2 43 54
DB032432.1 - 1 - 2 78 54

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300006462.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000009.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK290411.1,AL834397.1,BC005184.1,NM_001302946.1,NM_182916.2,XM_011533778.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRNT1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA 3'-terminal CCA addition
protein targeting to mitochondrion
tRNA 3'-end processing
mitochondrial tRNA 3'-end processing
tRNA binding
tRNA adenylyltransferase activity
ATP binding
RNA polymerase activity
CTP:tRNA cytidylyltransferase activity
CTP:3'-cytidine-tRNA cytidylyltransferase activity
ATP:3'-cytidine-cytidine-tRNA adenylyltransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000607052.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008990.hg.1 ENST00000438230.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008990.hg.1 ENST00000438230.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 199911 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0