This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01064353_m1
See other TRNT1 GE Assays ›
Gene Symbol:
TRNT1
Gene Name:
tRNA nucleotidyl transferase 1
Gene Aliases:
CCA1, CGI-47, MtCCA, RPEM, SIFD
Chromosome Location:
Chr.3: 3126916 - 3153435 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01064353_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRNT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC012537.1 - 8 - 9 1395 4097092 168

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300006462.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor nucleic acid metabolism protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000251607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000280591.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000339437.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000397779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000402675.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000413000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000420393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000465998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000469632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000482311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 ENST00000607052.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_001302946.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006462.hg.1 NM_182916.2 Refseq
Affymetrix Human Clariom D Assay PSR0300144326.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300144326.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300144326.hg.1 TC0300006462.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300144327.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300144327.hg.1 ENST00000434583.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300144327.hg.1 TC0300006462.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0