This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01064693_g1
See other GPR176 GE Assays ›
Gene Symbol:
GPR176
Gene Name:
G protein-coupled receptor 176
Gene Aliases:
HB-954
Chromosome Location:
Chr.15: 39795049 - 39920933 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01064693_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR176
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011521167.2 XP_011519469.1 1 - 2 496 132
XM_017021877.1 XP_016877366.1 1 - 2 479 132

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011521167.2, XM_017021877.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
chemical synaptic transmission
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0