This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01065219_m1
See other APPBP2 GE Assays ›
Gene Symbol:
APPBP2
Gene Name:
amyloid beta precursor protein binding protein 2
Gene Aliases:
APP-BP2, HS.84084, PAT1
Chromosome Location:
Chr.17: 60443149 - 60526240 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01065219_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APPBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006380.3 NP_006371.2 2 - 3 533 106
GenBank mRNA AF017782.1 - 2 - 3 509 106
AK292227.1 - 2 - 3 300 106
BC018121.1 - 2 - 3 436 3916891 106
D86981.1 - 2 - 3 512 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700011319.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001764.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APPBP2

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
intracellular transport
microtubule motor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17023555.hg.1 ENST00000083182.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17023555.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17023555.hg.1 TC17001764.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay PSR1700191260.hg.1 TC1700011319.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0