This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01065387_m1
See other GABPB1 GE Assays ›
Gene Symbol:
GABPB1
Gene Name:
GA binding protein transcription factor beta subunit 1
Gene Aliases:
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
Chromosome Location:
Chr.15: 50277192 - 50355429 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01065387_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GABPB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320910.1 NP_001307839.1 1 - 2 443 147
NM_001320915.1 NP_001307844.1 1 - 2 443 147
NM_181427.3 NP_852092.1 1 - 2 422 147
XM_017022053.1 XP_016877542.1 1 - 2 177 147
XM_017022054.1 XP_016877543.1 1 - 2 138 147
GenBank mRNA BQ430343.1 - 163 147
CB960428.1 - 238 147
D13316.1 - 1 - 2 115 145
DA502761.1 - 1 - 2 205 147

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the D13316.1,NM_001320910.1,NM_001320915.1,NM_181427.3,XM_017022053.1,XM_017022054.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GABPB1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
mitochondrion organization
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline