This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01065388_m1
See other GABPB1 GE Assays ›
Gene Symbol:
GABPB1
Gene Name:
GA binding protein transcription factor beta subunit 1
Gene Aliases:
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
Chromosome Location:
Chr.15: 50277192 - 50355429 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01065388_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GABPB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320910.1 NP_001307839.1 2 - 3 468 80
NM_001320915.1 NP_001307844.1 2 - 3 468 80
NM_181427.3 NP_852092.1 2 - 3 447 80
XM_017022053.1 XP_016877542.1 2 - 3 202 80
XM_017022054.1 XP_016877543.1 2 - 3 163 80
GenBank mRNA BQ430343.1 - 188 80
CB960428.1 - 263 80
D13316.1 - 2 - 3 140 80
DA502761.1 - 2 - 3 230 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the D13316.1, NM_001320910.1, NM_001320915.1, NM_181427.3, XM_017022053.1, XM_017022054.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GABPB1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
mitochondrion organization
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0