This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01065972_m1
See other N6AMT1 GE Assays ›
Gene Symbol:
N6AMT1
Gene Name:
N-6 adenine-specific DNA methyltransferase 1 (putative)
Gene Aliases:
C21orf127, HEMK2, MTQ2, N6AMT, PRED28, m.HsaHemK2P
Chromosome Location:
Chr.21: 28571095 - 28885373 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01065972_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N6AMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182749.4 NP_877426.4 3 - 4 342 83
GenBank mRNA BC011554.1 - 3 - 4 323 4640080 83
BP317563.1 - 3 - 4 339 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene N6AMT1

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

translational termination
protein methylation
positive regulation of cell growth
methylation
nucleic acid binding
protein binding
protein methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000303775.9 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000351429.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 ENST00000460212.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_013240.5 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NM_182749.4 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq
Affymetrix Human Clariom D Assay TC2100007846.hg.1 NR_047510.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0