This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01067927_g1
See other PPA1 GE Assays ›
Gene Symbol:
PPA1
Gene Name:
pyrophosphatase (inorganic) 1
Gene Aliases:
HEL-S-66p, IOPPP, PP, PP1, SID6-8061
Chromosome Location:
Chr.10: 70202830 - 70233434 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01067927_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021129.3 NP_066952.1 10 - 11 945 128
GenBank mRNA AB026723.1 - 10 - 11 868 128
AF092439.1 - 10 - 11 915 128
AF119665.1 - 10 - 11 915 128
AF154065.1 - 10 - 11 883 128
AF217186.1 - 10 - 11 838 128
AK058193.1 - 10 - 11 938 128
BC001022.2 - 10 - 11 920 3345496 128
BC061581.1 - 10 - 11 928 6526628 128
BC105034.1 - 10 - 11 911 8144037 128
BC105036.1 - 10 - 11 911 8144039 128
BC107882.1 - 10 - 11 879 6495107 128
EU794626.1 - 10 - 11 945 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010914.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001362.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPA1

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
phosphate-containing compound metabolic process
diphosphate metabolic process
magnesium ion binding
inorganic diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline