This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068014_m1
See other CHD9 GE Assays ›
Gene Symbol:
CHD9
Gene Name:
chromodomain helicase DNA binding protein 9
Gene Aliases:
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location:
Chr.16: 53054991 - 53327502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068014_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017023724.1 XP_016879213.1 3 - 4 1724 92
XM_017023725.1 XP_016879214.1 3 - 4 1724 92
XM_017023726.1 XP_016879215.1 1 - 2 931 92
GenBank mRNA AK299045.1 - 1 - 2 358 92
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF150735.1 - 3 - 4 Chr. 16 - 53207974 - 53226385 [-] on Build GRCh38 92
GenBankmRNA CN348885.1 - 1 - 2 349 Chr. 16 - 53207998 - 53222678 [+] on Build GRCh38 92
GenBankmRNA DC363484.1 - 1 - 2 362 Chr. 16 - 53207986 - 53209706 [+] on Build GRCh38 92
GenBankmRNA BE882430.1 - 308 Chr. - - [-] on Build 92
GenBankmRNA DA287640.1 - 1 - 2 315 Chr. 16 - 53208032 - 53222711 [+] on Build GRCh38 92
GenBankmRNA BU193734.1 - 1 - 2 260 Chr. 16 - 53208087 - 53226481 [+] on Build GRCh38 92
GenBankmRNA DC369270.1 - 1 - 2 358 Chr. 16 - 53207989 - 53209718 [+] on Build GRCh38 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream p53-regulated lncRNA 1 LOC105371267 PR-lncRNA-1 -155.1kb Anti-sense Chr. 16 - 53035690 - 53052873 on NCBI Build GRCh38
Downstream u3 small nucleolar ribonucleoprotein protein MPP10-like LOC643802 159.5kb Anti-sense Chr. 16 - 53365391 - 53371212 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK299045.1,XM_017023726.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
cellular lipid metabolic process
DNA binding
helicase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0