This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068036_m1
See other CHD9 GE Assays ›
Gene Symbol:
CHD9
Gene Name:
chromodomain helicase DNA binding protein 9
Gene Aliases:
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location:
Chr.16: 53054991 - 53327502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068036_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 27 - 28 5461 95
NM_025134.4 NP_079410.4 28 - 29 5506 95
XM_005256168.3 XP_005256225.1 28 - 29 5548 95
XM_005256169.2 XP_005256226.1 28 - 29 5570 95
XM_005256172.3 XP_005256229.1 28 - 29 5487 95
XM_005256176.3 XP_005256233.1 27 - 28 4155 95
XM_006721281.2 XP_006721344.1 28 - 29 5487 95
XM_006721283.3 XP_006721346.1 28 - 29 5708 95
XM_011523347.2 XP_011521649.1 29 - 30 5620 95
XM_017023718.1 XP_016879207.1 29 - 30 5743 95
XM_017023719.1 XP_016879208.1 30 - 31 5694 95
XM_017023720.1 XP_016879209.1 29 - 30 5588 95
XM_017023721.1 XP_016879210.1 29 - 30 5744 95
XM_017023722.1 XP_016879211.1 29 - 30 5744 95
XM_017023723.1 XP_016879212.1 29 - 30 5744 95
XM_017023724.1 XP_016879213.1 29 - 30 5570 95
XM_017023725.1 XP_016879214.1 29 - 30 5570 95
XM_017023726.1 XP_016879215.1 27 - 28 4777 95
XM_017023727.1 XP_016879216.1 27 - 28 4155 95
XM_017023728.1 XP_016879217.1 29 - 30 5604 95
GenBank mRNA AB002306.3 - 27 - 28 4929 95
AY243500.1 - 27 - 28 5384 95
AY243501.1 - 9 - 10 1066 95
BC140815.1 - 27 - 28 5338 9021683 95
BC144622.1 - 27 - 28 5338 9053153 95
DQ059482.1 - 18 - 19 2834 95
DQ333316.1 - 28 - 29 5600 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600007874.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000445.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD9

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
cellular lipid metabolic process
DNA binding
helicase activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16006200.hg.1 ENST00000219084.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16006200.hg.1 ENST00000398510.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16006200.hg.1 ENST00000447540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16006200.hg.1 ENST00000450543.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16006200.hg.1 NM_025134.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16006200.hg.1 TC16000445.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000219084.10 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000398510.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000447540.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000564845.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000566029.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000615216.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000622617.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 NM_001308319.1 Refseq
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 NM_025134.5 Refseq
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000219084.10 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000398510.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000447540.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000564845.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000566029.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000615216.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 ENST00000622617.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 NM_001308319.1 Refseq
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 NM_025134.5 Refseq
Affymetrix Human Clariom D Assay PSR1600157434.hg.1 TC1600007874.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0