This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068050_g1
See other CHD9 GE Assays ›
Gene Symbol:
CHD9
Gene Name:
chromodomain helicase DNA binding protein 9
Gene Aliases:
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location:
Chr.16: 53054991 - 53327502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068050_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 8 - 9 2538 141
NM_025134.4 NP_079410.4 9 - 10 2583 141
XM_005256168.3 XP_005256225.1 9 - 10 2625 141
XM_005256169.2 XP_005256226.1 9 - 10 2647 141
XM_005256172.3 XP_005256229.1 9 - 10 2564 141
XM_005256176.3 XP_005256233.1 8 - 9 1232 141
XM_006721281.2 XP_006721344.1 9 - 10 2564 141
XM_006721283.3 XP_006721346.1 9 - 10 2785 141
XM_011523347.2 XP_011521649.1 10 - 11 2697 141
XM_017023718.1 XP_016879207.1 10 - 11 2820 141
XM_017023719.1 XP_016879208.1 11 - 12 2771 141
XM_017023720.1 XP_016879209.1 10 - 11 2665 141
XM_017023721.1 XP_016879210.1 10 - 11 2821 141
XM_017023722.1 XP_016879211.1 10 - 11 2821 141
XM_017023723.1 XP_016879212.1 10 - 11 2821 141
XM_017023724.1 XP_016879213.1 10 - 11 2647 141
XM_017023725.1 XP_016879214.1 10 - 11 2647 141
XM_017023726.1 XP_016879215.1 8 - 9 1854 141
XM_017023727.1 XP_016879216.1 8 - 9 1232 141
XM_017023728.1 XP_016879217.1 10 - 11 2681 141
GenBank mRNA AB002306.3 - 8 - 9 2006 141
AK299045.1 - 8 - 9 1281 141
AY243500.1 - 8 - 9 2461 141
AY647157.1 - 9 - 10 2583 141
BC140815.1 - 8 - 9 2415 9021683 141
BC144622.1 - 8 - 9 2415 9053153 141
BX390832.2 - 427 141
DQ333316.1 - 9 - 10 2677 141

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD9

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
cellular lipid metabolic process
DNA binding
helicase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0