This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068618_m1
See other EPB41 GE Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068618_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166005.1 NP_001159477.1 14 - 15 2065 134
XM_005245753.1 XP_005245810.1 14 - 15 2140 134
XM_005245760.1 XP_005245817.1 14 - 15 2140 134
XM_005245761.1 XP_005245818.1 14 - 15 2140 134
XM_005245768.1 XP_005245825.1 14 - 15 2140 134
XM_005245772.4 XP_005245829.1 14 - 15 2140 134
XM_005245773.4 XP_005245830.1 14 - 15 2140 134
XM_006710434.1 XP_006710497.1 14 - 15 2140 134
XM_006710439.1 XP_006710502.1 14 - 15 2140 134
XM_011540956.1 XP_011539258.1 14 - 15 2140 134
XM_011540957.1 XP_011539259.1 14 - 15 2137 134
XM_011540958.1 XP_011539260.1 14 - 15 2140 134
XM_011540963.2 XP_011539265.1 14 - 15 2140 134
XM_011540964.1 XP_011539266.1 14 - 15 2036 134
XM_011540965.2 XP_011539267.1 14 - 15 2140 134
XM_017000581.1 XP_016856070.1 13 - 14 2035 134
XM_017000582.1 XP_016856071.1 13 - 14 2032 134
XM_017000584.1 XP_016856073.1 14 - 15 2431 134
XM_017000587.1 XP_016856076.1 14 - 15 2137 134
XM_017000588.1 XP_016856077.1 14 - 15 2137 134
XM_017000593.1 XP_016856082.1 14 - 15 2033 134
XM_017000598.1 XP_016856087.1 14 - 15 2036 134
XM_017000599.1 XP_016856088.1 14 - 15 2036 134
XM_017000600.1 XP_016856089.1 14 - 15 2036 134
XM_017000601.1 XP_016856090.1 14 - 15 2033 134
GenBank mRNA AF156225.1 - 13 - 14 1938 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0