This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068711_g1
See other CEL GE Assays ›
Gene Symbol:
CEL
Gene Name:
carboxyl ester lipase
Gene Aliases:
BAL, BSDL, BSSL, CELL, CEase, FAP, FAPP, LIPA, MODY8
Chromosome Location:
Chr.9: 133061978 - 133071863 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068711_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001807.4 NP_001798.2 6 - 7 805 74
GenBank mRNA AF081673.1 - 5 - 6 720 74
AK299965.1 - 6 - 7 682 74
BC042510.1 - 6 - 7 790 5187959 74
M85201.1 - 6 - 7 797 74
S79774.1 - 6 - 7 799 74
X54457.1 - 6 - 7 870 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900012185.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000767.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CEL

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride metabolic process
cholesterol catabolic process
neuron cell-cell adhesion
synapse assembly
fatty acid catabolic process
protein esterification
pancreatic juice secretion
intestinal cholesterol absorption
lipid digestion
intestinal lipid catabolic process
modulation of synaptic transmission
catalytic activity
sterol esterase activity
triglyceride lipase activity
receptor activity
protein binding
heparin binding
hydrolase activity
neurexin family protein binding
acylglycerol lipase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09009860.hg.1 TC09000767.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09009860.hg.1 ENST00000303626.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009860.hg.1 ENST00000351304.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009860.hg.1 ENST00000372080.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09009860.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay TC0900012185.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay PSR0900185129.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900185129.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay PSR0900185129.hg.1 ENST00000372080.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900185129.hg.1 NM_001807.4 Refseq
Affymetrix Human Clariom D Assay PSR0900185129.hg.1 TC0900012185.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0