This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01068731_m1
See other RBPMS GE Assays ›
Gene Symbol:
RBPMS
Gene Name:
RNA binding protein with multiple splicing
Gene Aliases:
HERMES
Chromosome Location:
Chr.8: 30384501 - 30572261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01068731_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBPMS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008710.2 NP_001008710.1 7 - 8 1191 113
GenBank mRNA D84107.1 - 7 - 8 1165 113
D84111.1 - 6 - 7 611 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBPMS

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
response to oxidative stress
positive regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of SMAD protein import into nucleus
nucleotide binding
transcription coactivator activity
RNA binding
protein binding
poly(A) binding
snRNA binding
protein homodimerization activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline