This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01070658_m1
See other CRIM1 GE Assays ›
Gene Symbol:
CRIM1
Gene Name:
cysteine rich transmembrane BMP regulator 1
Gene Aliases:
CRIM-1, S52
Chromosome Location:
Chr.2: 36356088 - 36551135 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01070658_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRIM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016441.2 NP_057525.1 11 - 12 2058 74
XM_005264357.4 XP_005264414.1 10 - 11 1880 74
XM_011532898.2 XP_011531200.1 12 - 13 2179 74
XM_011532899.2 XP_011531201.1 12 - 13 2180 74
XM_011532901.2 XP_011531203.1 12 - 13 2180 74
XM_017004258.1 XP_016859747.1 11 - 12 1936 74
XM_017004259.1 XP_016859748.1 11 - 12 2197 74
XM_017004260.1 XP_016859749.1 10 - 11 1811 74
XM_017004261.1 XP_016859750.1 10 - 11 1471 74
XM_017004262.1 XP_016859751.1 9 - 10 1354 74
GenBank mRNA AB209139.1 - 11 - 12 2058 74
AF167706.1 - 11 - 12 2031 74
AY358371.1 - 11 - 12 2359 74
BC111989.1 - 11 - 12 2029 8327457 74
BC113371.1 - 11 - 12 2029 8322423 74
BX648755.1 - 5 - 6 707 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200007237.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000230.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CRIM1

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
nervous system development
negative regulation of endopeptidase activity
insulin-like growth factor receptor signaling pathway
serine-type endopeptidase inhibitor activity
insulin-like growth factor-activated receptor activity
insulin-like growth factor binding
PDZ domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02003408.hg.1 ENST00000280527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02003408.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02003408.hg.1 TC02000230.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02003407.hg.1 ENST00000280527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02003407.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02003407.hg.1 TC02000230.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000473403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000473403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000473403.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000477491.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000477491.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000477491.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000481321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000481321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000481321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000497236.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 ENST00000497236.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay PSR0200150098.hg.1 TC0200007237.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000426856.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000428774.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000473403.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000473403.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000473403.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000477491.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000477491.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000477491.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000481321.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000481321.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000481321.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000497236.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 ENST00000497236.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay TC0200007237.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 ENST00000280527.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 ENST00000413985.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 NM_016441.2 Refseq
Affymetrix Human Clariom D Assay PSR0200150099.hg.1 TC0200007237.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline