This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01071257_m1
See other MOGAT1 GE Assays ›
Gene Symbol:
MOGAT1
Gene Name:
monoacylglycerol O-acyltransferase 1
Gene Aliases:
DGAT2L, DGAT2L1, MGAT1
Chromosome Location:
Chr.2: 222671738 - 222709930 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01071257_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOGAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_058165.2 NP_477513.2 2 - 3 317 156
GenBank mRNA AF384163.1 - 2 - 3 314 156

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010902.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001349.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MOGAT1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
diacylglycerol biosynthetic process
triglyceride biosynthetic process
2-acylglycerol O-acyltransferase activity
diacylglycerol O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02021038.hg.1 TC02001349.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02021038.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02021038.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay PSR0200178155.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200178155.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay PSR0200178155.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200178155.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay PSR0200178155.hg.1 TC0200010902.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0