This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01071560_m1
See other NECTIN2 GE Assays ›
Gene Symbol:
NECTIN2
Gene Name:
nectin cell adhesion molecule 2
Gene Aliases:
CD112, HVEB, PRR2, PVRL2, PVRR2
Chromosome Location:
Chr.19: 44846136 - 44889228 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01071560_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NECTIN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042724.1 NP_001036189.1 1 - 2 480 101
NM_002856.2 NP_002847.1 1 - 2 480 101
XM_011527192.2 XP_011525494.1 1 - 2 480 101
GenBank mRNA AF058448.1 - 1 - 2 239 101
AK291330.1 - 1 - 2 306 101
BC003091.1 - 1 - 2 288 3503222 101
CN410857.1 - 1 - 2 452 101
CR456818.1 - 1 - 2 90 101
X80038.1 - 1 - 2 90 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008287.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000631.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NECTIN2

Gene Ontology Categories:

Function(s) Process(es)

acrosome assembly
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cytoskeleton organization
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
spermatid development
spermatid nucleus differentiation
cell recognition
fertilization
fusion of virus membrane with host plasma membrane
sperm mitochondrion organization
cell part morphogenesis
positive regulation of mast cell activation
adherens junction organization
susceptibility to natural killer cell mediated cytotoxicity
adhesion of symbiont to host
cilium organization
positive regulation of natural killer cell mediated cytotoxicity
regulation of viral entry into host cell
coreceptor-mediated virion attachment to host cell
regulation of immune response
establishment of mitochondrion localization
susceptibility to T cell mediated cytotoxicity
virus receptor activity
receptor binding
protein binding
coreceptor activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19008759.hg.1 ENST00000252483.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008759.hg.1 ENST00000252485.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008759.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008759.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008759.hg.1 TC19000631.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000592018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000592018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 ENST00000592018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008760.hg.1 ENST00000252483.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008760.hg.1 ENST00000252485.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008760.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008760.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008760.hg.1 TC19000631.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19008761.hg.1 ENST00000252483.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008761.hg.1 ENST00000252485.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008761.hg.1 NM_001042724.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008761.hg.1 NM_002856.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008761.hg.1 TC19000631.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 TC1900008287.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163321.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 TC1900008287.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163319.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163316.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163316.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163316.hg.1 TC1900008287.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900163316.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163316.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 TC1900008287.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163317.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 TC1900008287.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay PSR1900163318.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252483.9 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000252485.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000585601.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000587386.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000591581.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000592018.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000592018.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 ENST00000592018.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_001042724.1 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq
Affymetrix Human Clariom D Assay TC1900008287.hg.1 NM_002856.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0