This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01072552_s1
See other HIST1H3I GE Assays ›
Gene Symbol:
HIST1H3I
Gene Name:
histone cluster 1, H3i
Gene Aliases:
H3.f, H3/f, H3FF
Chromosome Location:
Chr.6: 27871845 - 27872321 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01072552_s1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIST1H3I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003533.2 NP_003524.1 1 433 117
GenBank mRNA BC067493.1 - 1 433 7002079 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011233.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001427.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HIST1H3I

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
positive regulation of defense response to virus by host
nucleosome assembly
DNA replication-dependent nucleosome assembly
blood coagulation
gene silencing by RNA
telomere organization
cellular protein metabolic process
negative regulation of gene expression, epigenetic
positive regulation of gene expression, epigenetic
protein heterotetramerization
regulation of gene silencing
cell-cell adhesion
xenophagy
DNA binding
protein binding
histone binding
protein heterodimerization activity
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P24820499 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P24820501 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P04630312 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P04630313 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P04630314 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 29663 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 20354 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 62118 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 34651 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 516893 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P9321 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P9321 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 21076 - -
Agilent Human Genome CGH Microarray 44B A_14_P105939 - -
Agilent Human Genome CGH Microarray 44A A_14_P105939 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26389 - -
Agilent Human Genome CGH + SNP Microarray A_18_P15586447 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P9321 - -
SurePrint G3 Human GE Microarray G4851A A_24_P9321 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820499 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630314 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820502 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820499 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24820502 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630314 - -
Whole Human Genome Oligo Microarray G4112A A_24_P9321 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P105939 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P04630314 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P04630318 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P04630316 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P04630320 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P04630313 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04630312 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04630313 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04630314 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P105939 - -
Agilent Human Genome CGH Microarray 4x44K A_14_P105939 - -
Agilent Human Genome CGH Microarray 4x44K A_14_P111746 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P9321 - -
Custom Human Agilent Array 11k, Chromosome 6, Expression 815 - -
Custom Human Agilent Array 11k, Chromosome 6, Expression 1166 - -
Custom Human Agilent Array 11k, Chromosome 6, Expression 2922 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 84645 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 14937 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P111746 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P105939 - -
Agilent Human 1B Microarray G4111A A_32_P806675 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630324 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630269 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630318 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15796527 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630272 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630278 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630260 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630333 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630343 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630287 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630283 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630258 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630335 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630338 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630271 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630262 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630326 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630316 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630254 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15796534 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630320 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630291 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630313 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630250 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630285 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630276 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630328 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630267 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630331 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630341 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630339 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630248 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P04630251 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P105939 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P111746 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_003533_at - -
Affymetrix Human Gene 1.0 ST Array 8124532 - -
Affymetrix Human Gene 1.0 ST Array 8124533 - -
Affymetrix Human Gene 1.0 ST Array 8117417 - -
Affymetrix Human Gene 1.0 ST Array 8117416 - -
Affymetrix Human Gene 1.1 ST Array 8124531 - -
Affymetrix HT HG-U133+ PM Array Plate 214509_PM_at - -
Affymetrix Human Genome U219 Array 11733426_PM_at - -
Affymetrix Human Genome U219 Array 11733426_at - -
Affymetrix Human Transcriptome Array 2.0 PSR06016360.hg.1 TC06001427.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06016360.hg.1 ENST00000328488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06016360.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175735.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175735.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175735.hg.1 TC0600011233.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 214509_at - -
Human Genome U133 Plus 2.0 Array 214472_PM_at - -
Human Genome U133 Plus 2.0 Array 208575_at - -
Human Genome U133 Plus 2.0 Array 208496_PM_x_at - -
Human Genome U133 Plus 2.0 Array 214616_PM_at - -
Human Genome U133 Plus 2.0 Array 214472_at - -
Human Genome U133 Plus 2.0 Array 208496_x_at - -
Human Genome U133 Plus 2.0 Array 214509_PM_at - -
Human Genome U133 Plus 2.0 Array 208577_PM_at - -
Human Genome U133 Plus 2.0 Array 208577_at - -
Human Genome U133 Plus 2.0 Array 208576_PM_s_at - -
Human Genome U133 Plus 2.0 Array 214616_at - -
Human Genome U133 Plus 2.0 Array 208575_PM_at - -
Human Genome U133 Plus 2.0 Array 208506_PM_at - -
Human Genome U133 Plus 2.0 Array 208576_s_at - -
Human Genome U133 Plus 2.0 Array 208506_at - -
Human Genome U133A 2.0 Array 214509_PM_at - -
Human Genome U133A 2.0 Array 208576_s_at - -
Human Genome U133A 2.0 Array 214509_at - -
Human Genome U133A 2.0 Array 208577_at - -
Human Genome U133A 2.0 Array 214472_at - -
Human Genome U133A 2.0 Array 208506_PM_at - -
Human Genome U133A 2.0 Array 208496_x_at - -
Human Genome U133A 2.0 Array 208496_PM_x_at - -
Human Genome U133A 2.0 Array 208575_at - -
Human Genome U133A 2.0 Array 208506_at - -
Human Genome U133A 2.0 Array 214616_PM_at - -
Human Genome U133A 2.0 Array 208577_PM_at - -
Human Genome U133A 2.0 Array 214472_PM_at - -
Human Genome U133A 2.0 Array 208576_PM_s_at - -
Human Genome U133A 2.0 Array 214616_at - -
Human Genome U133A 2.0 Array 208575_PM_at - -
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_003533_at - -
AffymetrixPathwork Diagnostics Pathchip pw590173 - -
Human Genome U95 Set 35598_at - -
Human Genome U95 Set 33018_at - -
Human Genome U95 Set 35598_PM_at - -
Human Genome U95 Set 33018_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 214509_at5 - -
Human Genome U133 Set 208575_PM_at - -
Human Genome U133 Set 208576_PM_s_at - -
Human Genome U133 Set 208576_s_at - -
Human Genome U133 Set 208575_at - -
Human Genome U133 Set 208506_PM_at - -
Human Genome U133 Set 208506_at - -
Human Genome U133 Set 214616_PM_at - -
Human Genome U133 Set 208577_at - -
Human Genome U133 Set 214472_PM_at - -
Human Genome U133 Set 214509_PM_at - -
Human Genome U133 Set 208577_PM_at - -
Human Genome U133 Set 214472_at - -
Human Genome U133 Set 208496_PM_x_at - -
Human Genome U133 Set 214509_at - -
Human Genome U133 Set 214616_at - -
Human Genome U133 Set 208496_x_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A029573 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A029573 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 174536 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_2207865 BC067493.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2207865 BC067494.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2207865 BC069305.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2207865 NM_003533.2 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2207865 BC066884.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1742688 - -
HumanHT-12 V4.0 expression beadchip ILMN_2207865 BC067493.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2207865 BC067494.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2207865 BC069305.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2207865 NM_003533.2 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2207865 BC066884.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2207865 BC067493.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2207865 BC067494.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2207865 BC069305.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2207865 NM_003533.2 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2207865 BC066884.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2207865 BC067493.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2207865 BC067494.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2207865 BC069305.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2207865 NM_003533.2 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2207865 BC066884.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_15718723-S BC067493.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_15718723-S BC067494.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_15718723-S BC069305.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_15718723-S NM_003533.2 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_15718723-S BC066884.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_29010-S - -
Illumina human-6 v1.0 expression beadchip 0006620279 - -
Illumina humanRef-8 v1.0 expression beadchip 0006620279 - -
Illumina human-6 v2.0 expression beadchip ILMN_1742688 - -
Illumina humanRef-8 v2.0 expression beadchip ILMN_1742688 - -
Illumina Sentrix Human-6 v2 Expression BeadChip 6560706 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_29010 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2207865 BC067493.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2207865 BC067494.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2207865 BC069305.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2207865 NM_003533.2 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2207865 BC066884.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2207865 BC067493.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2207865 BC067494.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2207865 BC069305.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2207865 NM_003533.2 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2207865 BC066884.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2207865 BC067493.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2207865 BC067494.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2207865 BC069305.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2207865 NM_003533.2 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2207865 BC066884.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1742688 - -
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2207865 BC067493.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2207865 BC067494.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2207865 BC069305.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2207865 NM_003533.2 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2207865 BC066884.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0