This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01073650_m1
See other GTF2I GE Assays ›
Gene Symbol:
GTF2I
Gene Name:
general transcription factor IIi
Gene Aliases:
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6
Chromosome Location:
Chr.7: 74657665 - 74760692 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01073650_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163636.2 NP_001157108.1 10 - 11 1231 91
NM_032999.3 NP_127492.1 10 - 11 1234 91
NM_033001.3 NP_127494.1 10 - 11 1234 91
GenBank mRNA AF035737.1 - 10 - 11 1141 91
AF038968.1 - 10 - 11 1141 91
AK294935.1 - 11 - 12 1068 91
BC099907.1 - 10 - 11 1163 5263270 91
KJ534860.1 - 9 - 10 825 91
KJ534862.1 - 9 - 10 825 91
KJ534863.1 - 9 - 10 825 91
KJ534965.1 - 9 - 10 825 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2I

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
signal transduction
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0