This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01074095_m1
See other XPO4 GE Assays ›
Gene Symbol:
XPO4
Gene Name:
exportin 4
Gene Aliases:
exp4
Chromosome Location:
Chr.13: 20777329 - 20902774 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01074095_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XPO4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022459.4 NP_071904.4 16 - 17 2358 105
XM_005266502.1 XP_005266559.1 17 - 18 2513 105
XM_005266503.3 XP_005266560.1 16 - 17 2358 105
XM_005266504.3 XP_005266561.1 10 - 11 1885 105
XM_011535197.1 XP_011533499.1 17 - 18 2522 105
XM_011535198.1 XP_011533500.1 15 - 16 2200 105
XM_011535199.1 XP_011533501.1 14 - 15 2163 105
XM_011535200.2 XP_011533502.1 14 - 15 3613 105
XM_017020704.1 XP_016876193.1 12 - 13 2154 105
XM_017020705.1 XP_016876194.1 13 - 14 3390 105
XM_017020706.1 XP_016876195.1 10 - 11 1509 105
GenBank mRNA AB051508.1 - 16 - 17 2321 105
AK023108.1 - 8 - 9 1314 105
AK057924.1 - 16 - 17 2328 105
BC042504.1 - 13 - 14 1894 105
BC050680.1 - 13 - 14 1911 105
BC113571.1 - 15 - 16 2261 8322623 105
BC113573.1 - 15 - 16 2261 8322625 105
BX647843.1 - 7 - 8 1129 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008267.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000461.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene XPO4

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein export from nucleus
positive regulation of protein export from nucleus
nuclear export signal receptor activity
protein binding
Ran GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005253.hg.1 ENST00000255305.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005253.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005253.hg.1 ENST00000456108.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005253.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005253.hg.1 TC13000461.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155809.hg.1 TC1300008267.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline