This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01074096_g1
See other XPO4 GE Assays ›
Gene Symbol:
XPO4
Gene Name:
exportin 4
Gene Aliases:
exp4
Chromosome Location:
Chr.13: 20777329 - 20902774 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01074096_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XPO4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022459.4 NP_071904.4 17 - 18 2657 122
XM_005266502.1 XP_005266559.1 18 - 19 2812 122
XM_005266503.3 XP_005266560.1 17 - 18 2657 122
XM_005266504.3 XP_005266561.1 11 - 12 2184 122
XM_011535197.1 XP_011533499.1 18 - 19 2821 122
XM_011535198.1 XP_011533500.1 16 - 17 2499 122
XM_011535199.1 XP_011533501.1 15 - 16 2462 122
XM_011535200.2 XP_011533502.1 15 - 16 3912 122
XM_017020704.1 XP_016876193.1 13 - 14 2453 122
XM_017020705.1 XP_016876194.1 14 - 15 3689 122
XM_017020706.1 XP_016876195.1 11 - 12 1808 122
GenBank mRNA AB051508.1 - 17 - 18 2620 122
AK023108.1 - 9 - 10 1613 122
AK057924.1 - 17 - 18 2627 122
BC042504.1 - 14 - 15 2193 122
BC050680.1 - 14 - 15 2210 122
BC113571.1 - 16 - 17 2560 8322623 122
BC113573.1 - 16 - 17 2560 8322625 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008267.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000461.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene XPO4

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein export from nucleus
positive regulation of protein export from nucleus
nuclear export signal receptor activity
protein binding
Ran GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005252.hg.1 ENST00000255305.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005252.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005252.hg.1 ENST00000456108.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005252.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005252.hg.1 TC13000461.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155808.hg.1 TC1300008267.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline