Assay Details
| Gene Symbol: XPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022459.4 | NP_071904.4 | 6 - 7 | 769 | 93 | |
| XM_005266502.1 | XP_005266559.1 | 7 - 8 | 924 | 93 | ||
| XM_005266503.3 | XP_005266560.1 | 6 - 7 | 769 | 93 | ||
| XM_011535197.1 | XP_011533499.1 | 7 - 8 | 933 | 93 | ||
| XM_011535198.1 | XP_011533500.1 | 5 - 6 | 611 | 93 | ||
| XM_011535199.1 | XP_011533501.1 | 4 - 5 | 574 | 93 | ||
| XM_011535200.2 | XP_011533502.1 | 4 - 5 | 2024 | 93 | ||
| XM_017020704.1 | XP_016876193.1 | 2 - 3 | 565 | 93 | ||
| XM_017020705.1 | XP_016876194.1 | 3 - 4 | 1801 | 93 | ||
| GenBank mRNA | AB051508.1 | - | 6 - 7 | 732 | 93 | |
| AK057924.1 | - | 6 - 7 | 739 | 93 | ||
| BC042504.1 | - | 3 - 4 | 305 | 93 | ||
| BC050680.1 | - | 3 - 4 | 322 | 93 | ||
| BC113571.1 | - | 5 - 6 | 672 | 8322623 | 93 | |
| BC113573.1 | - | 5 - 6 | 672 | 8322625 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000461.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005263.hg.1 | ENST00000255305.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005263.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005263.hg.1 | ENST00000456108.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005263.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005263.hg.1 | TC13000461.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155819.hg.1 | TC1300008267.hg.1 | Clariom D TC |

XPO4
Genomic Map