This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01074108_m1
See other XPO4 GE Assays ›
Gene Symbol:
XPO4
Gene Name:
exportin 4
Gene Aliases:
exp4
Chromosome Location:
Chr.13: 20777329 - 20902774 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01074108_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XPO4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022459.4 NP_071904.4 7 - 8 871 100
XM_005266502.1 XP_005266559.1 8 - 9 1026 100
XM_005266503.3 XP_005266560.1 7 - 8 871 100
XM_011535197.1 XP_011533499.1 8 - 9 1035 100
XM_011535198.1 XP_011533500.1 6 - 7 713 100
XM_011535199.1 XP_011533501.1 5 - 6 676 100
XM_011535200.2 XP_011533502.1 5 - 6 2126 100
XM_017020704.1 XP_016876193.1 3 - 4 667 100
XM_017020705.1 XP_016876194.1 4 - 5 1903 100
GenBank mRNA AB051508.1 - 7 - 8 834 100
AK057924.1 - 7 - 8 841 100
BC042504.1 - 4 - 5 407 100
BC050680.1 - 4 - 5 424 100
BC113571.1 - 6 - 7 774 8322623 100
BC113573.1 - 6 - 7 774 8322625 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008267.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000461.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene XPO4

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein export from nucleus
positive regulation of protein export from nucleus
nuclear export signal receptor activity
protein binding
Ran GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13005262.hg.1 ENST00000255305.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005262.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005262.hg.1 ENST00000456108.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005262.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005262.hg.1 TC13000461.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000465018.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 ENST00000490513.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay TC1300008267.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 ENST00000255305.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 ENST00000400602.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 NM_022459.4 Refseq
Affymetrix Human Clariom D Assay PSR1300155818.hg.1 TC1300008267.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline