This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01074403_m1
See other LTN1 GE Assays ›
Gene Symbol:
LTN1
Gene Name:
listerin E3 ubiquitin protein ligase 1
Gene Aliases:
C21orf10, C21orf98, RNF160, ZNF294
Chromosome Location:
Chr.21: 28928144 - 28992959 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01074403_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LTN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320766.1 NP_001307695.1 23 - 24 4409 152
NM_015565.2 NP_056380.2 24 - 25 4448 152
XM_017028316.1 XP_016883805.1 23 - 24 4128 152
XM_017028317.1 XP_016883806.1 14 - 15 2723 152
GenBank mRNA AB018257.2 - 24 - 25 4339 152
BC140790.1 - 24 - 25 4361 9021658 152
BC150284.1 - 24 - 25 4339 8860138 152
CN361594.1 - 4 - 5 565 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007850.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000351.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LTN1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein autoubiquitination
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21003979.hg.1 ENST00000361371.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21003979.hg.1 ENST00000389194.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21003979.hg.1 ENST00000446611.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21003979.hg.1 NM_015565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21003979.hg.1 TC21000351.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 ENST00000361371.9 Ensembl
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 ENST00000389194.6 Ensembl
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 ENST00000614971.4 Ensembl
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 NM_015565.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 TC2100007850.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 ENST00000361371.9 Ensembl
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 ENST00000389194.6 Ensembl
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 ENST00000614971.4 Ensembl
Affymetrix Human Clariom D Assay PSR2100152909.hg.1 NM_015565.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rzthw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0