This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01075232_m1
See other STARD5 GE Assays ›
Gene Symbol:
STARD5
Gene Name:
StAR related lipid transfer domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.15: 81312666 - 81324183 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01075232_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181900.2 NP_871629.1 1 - 2 182 58
GenBank mRNA AF480304.1 - 1 - 2 98 58
BC004365.1 - 1 - 2 124 3641640 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500010909.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001730.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STARD5

Gene Ontology Categories:

Function(s) Process(es)

C21-steroid hormone biosynthetic process
lipid transport
cholesterol binding
bile acid binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15016923.hg.1 ENST00000302824.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15016923.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15016923.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15016923.hg.1 TC15001730.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15016921.hg.1 ENST00000302824.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15016921.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15016921.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15016921.hg.1 TC15001730.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15016925.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15016925.hg.1 TC15001730.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185718.hg.1 TC1500010909.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500185720.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185720.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185720.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185720.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185720.hg.1 TC1500010909.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185714.hg.1 TC1500010909.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185717.hg.1 TC1500010909.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 210466 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0