This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01075233_g1
See other STARD5 GE Assays ›
Gene Symbol:
STARD5
Gene Name:
StAR related lipid transfer domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.15: 81312666 - 81324183 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01075233_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181900.2 NP_871629.1 2 - 3 233 72
XM_006720703.1 XP_006720766.1 2 - 3 390 72
GenBank mRNA AF480304.1 - 2 - 3 149 72
BC004365.1 - 2 - 3 175 3641640 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500010909.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001730.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STARD5

Gene Ontology Categories:

Function(s) Process(es)

C21-steroid hormone biosynthetic process
lipid transport
cholesterol binding
bile acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15016920.hg.1 ENST00000302824.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15016920.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15016920.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15016920.hg.1 TC15001730.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay PSR1500185712.hg.1 TC1500010909.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0