Assay Details
| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042724.1 | NP_001036189.1 | 7 - 8 | 1648 | 154 | |
| GenBank mRNA | X80038.1 | - | 7 - 8 | 1258 | 154 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000631.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
acrosome assembly
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cytoskeleton organization
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
spermatid development
spermatid nucleus differentiation
cell recognition
fertilization
fusion of virus membrane with host plasma membrane
sperm mitochondrion organization
cell part morphogenesis
positive regulation of mast cell activation
adherens junction organization
susceptibility to natural killer cell mediated cytotoxicity
adhesion of symbiont to host
cilium organization
positive regulation of natural killer cell mediated cytotoxicity
regulation of viral entry into host cell
coreceptor-mediated virion attachment to host cell
regulation of immune response
establishment of mitochondrion localization
susceptibility to T cell mediated cytotoxicity
virus receptor activity
receptor binding
protein binding
coreceptor activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
cell adhesion molecule binding
acrosome assembly
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cytoskeleton organization
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
spermatid development
spermatid nucleus differentiation
cell recognition
fertilization
fusion of virus membrane with host plasma membrane
sperm mitochondrion organization
cell part morphogenesis
positive regulation of mast cell activation
adherens junction organization
susceptibility to natural killer cell mediated cytotoxicity
adhesion of symbiont to host
cilium organization
positive regulation of natural killer cell mediated cytotoxicity
regulation of viral entry into host cell
coreceptor-mediated virion attachment to host cell
regulation of immune response
establishment of mitochondrion localization
susceptibility to T cell mediated cytotoxicity
virus receptor activity
receptor binding
protein binding
coreceptor activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
cell adhesion molecule binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19008769.hg.1 | ENST00000252483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008769.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008769.hg.1 | TC19000631.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008770.hg.1 | ENST00000252483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008770.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008770.hg.1 | TC19000631.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011978.hg.1 | ENST00000462510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000252485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000585601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000587386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000591581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008287.hg.1 | NM_002856.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | TC1900008287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | ENST00000592018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163336.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011978.hg.1 | ENST00000462510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163338.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163338.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163338.hg.1 | TC1900008287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163338.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163338.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163337.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163337.hg.1 | NM_001042724.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163337.hg.1 | TC1900008287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163337.hg.1 | ENST00000252483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163337.hg.1 | NM_001042724.1 | Refseq |

NECTIN2
Genomic Map