This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01075804_m1
See other ADSL GE Assays ›
Gene Symbol:
ADSL
Gene Name:
adenylosuccinate lyase
Gene Aliases:
AMPS, ASASE, ASL
Chromosome Location:
Chr.22: 40346500 - 40387408 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01075804_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADSL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017028636.1 XP_016884125.1 2 - 3 400 106
XM_017028637.1 XP_016884126.1 2 - 3 383 106

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide biosynthetic process
AMP biosynthetic process
'de novo' IMP biosynthetic process
purine ribonucleoside monophosphate biosynthetic process
'de novo' AMP biosynthetic process
protein tetramerization
N6-(1,2-dicarboxyethyl)AMP AMP-lyase (fumarate-forming) activity
(S)-2-(5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamido)succinate AMP-lyase (fumarate-forming) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bjcbn:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0