This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01076124_m1
See other ATOX1 GE Assays ›
Gene Symbol:
ATOX1
Gene Name:
antioxidant 1 copper chaperone
Gene Aliases:
ATX1, HAH1
Chromosome Location:
Chr.5: 151742822 - 151758649 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01076124_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004045.3 NP_004036.1 1 - 2 102 91
GenBank mRNA AK293063.1 - 1 - 2 109 91
BC112248.1 - 1 - 2 184 8327716 91
BC112250.1 - 1 - 2 184 8327718 91
BI599233.1 - 1 - 2 110 91
BU172675.1 - 1 - 2 480 91
CN280769.1 - 1 - 2 79 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012523.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001957.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK293063.1,BC112248.1,BC112250.1,NM_004045.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATOX1

Gene Ontology Categories:

Function(s) Process(es)

copper ion transport
cellular copper ion homeostasis
response to oxidative stress
intracellular copper ion transport
copper ion transmembrane transport
copper ion transmembrane transporter activity
copper ion binding
protein binding
metallochaperone activity
copper chaperone activity
copper-dependent protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05027273.hg.1 TC05001957.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05027273.hg.1 ENST00000313115.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027273.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027273.hg.1 ENST00000524142.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027273.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027272.hg.1 TC05001957.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05027272.hg.1 ENST00000313115.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027272.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027272.hg.1 ENST00000524142.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027272.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 TC05001957.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 ENST00000313115.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 ENST00000520382.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 ENST00000521264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 ENST00000522314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 ENST00000524142.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027267.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187422.hg.1 TC0500012523.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522145.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay TC0500012523.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000520382.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000521264.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000522314.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187415.hg.1 TC0500012523.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 ENST00000313115.10 Ensembl
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 ENST00000522710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 NM_004045.3 Refseq
Affymetrix Human Clariom D Assay PSR0500187421.hg.1 TC0500012523.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500187424.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187424.hg.1 ENST00000524142.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187424.hg.1 TC0500012523.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 169363 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0