This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01076250_g1
See other TMCO6 GE Assays ›
Gene Symbol:
TMCO6
Gene Name:
transmembrane and coiled-coil domains 6
Gene Aliases:
PRO1580
Chromosome Location:
Chr.5: 140596529 - 140647411 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01076250_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMCO6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300980.1 NP_001287909.1 6 - 7 794 68
XM_011537663.2 XP_011535965.1 6 - 7 794 68
XM_011537665.2 XP_011535967.1 5 - 6 858 68
XM_017009618.1 XP_016865107.1 6 - 7 794 68
XM_017009619.1 XP_016865108.1 5 - 6 582 68
GenBank mRNA BC001910.1 - 6 - 7 717 3538001 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CV806910.1 - 2 - 3 168 Chr. 5 - 140642342 - 140644677 [+] on Build GRCh38 68
GenBankmRNA CD513649.1 - 3 - 4 351 Chr. 5 - 140641898 - 140644153 [+] on Build GRCh38 68
GenBankmRNA AF119848.1 - Chr. - - [-] on Build 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CD14 molecule CD14 -8.9kb Anti-sense Chr. 5 - 140631728 - 140633701 on NCBI Build GRCh38
Downstream NADH:ubiquinone oxidoreductase subunit A2 NDUFA2 B8,
CD14,
CIB8
2.3kb Anti-sense Chr. 5 - 140645363 - 140647785 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC001910.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus
protein transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0