This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01076902_m1
See other CLNK GE Assays ›
Gene Symbol:
CLNK
Gene Name:
cytokine dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.4: 10489899 - 10734853 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01076902_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_052964.2 NP_443196.2 16 - 17 1049 76
XM_011513775.2 XP_011512077.1 16 - 17 1280 76
XM_017007684.1 XP_016863173.1 16 - 17 1280 76
XM_017007685.1 XP_016863174.1 16 - 17 1280 76
GenBank mRNA AB032369.1 - 14 - 15 858 76
AB110420.1 - 16 - 17 1152 76
AK093001.1 - 16 - 17 1051 76
BC137553.1 - 16 - 17 9021553 76
BC137556.1 - 16 - 17 978 9021556 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010050.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001036.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLNK

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of signal transduction
intracellular signal transduction
SH3/SH2 adaptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04014215.hg.1 ENST00000226951.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014215.hg.1 ENST00000429087.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014215.hg.1 ENST00000515667.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014215.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014215.hg.1 TC04001036.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168916.hg.1 TC0400010050.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline