This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01076905_m1
See other CLNK GE Assays ›
Gene Symbol:
CLNK
Gene Name:
cytokine dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.4: 10489899 - 10734853 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01076905_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC029887.1 - 1 - 2 56 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC029887.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline