This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01076907_m1
See other CLNK GE Assays ›
Gene Symbol:
CLNK
Gene Name:
cytokine dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.4: 10489899 - 10734853 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01076907_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_052964.2 NP_443196.2 4 - 5 250 94
XM_011513775.2 XP_011512077.1 4 - 5 481 94
XM_017007684.1 XP_016863173.1 4 - 5 481 94
XM_017007685.1 XP_016863174.1 4 - 5 481 94
GenBank mRNA AB032369.1 - 2 - 3 59 94
AB110420.1 - 4 - 5 353 94
AK093001.1 - 4 - 5 252 94
BC029887.1 - 4 - 5 280 94
BC137553.1 - 4 - 5 179 9021553 94
BC137556.1 - 4 - 5 179 9021556 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010050.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001036.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC029887.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLNK

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of signal transduction
intracellular signal transduction
SH3/SH2 adaptor activity
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04014237.hg.1 ENST00000226951.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014237.hg.1 ENST00000429087.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014237.hg.1 ENST00000442825.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014237.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014237.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014237.hg.1 TC04001036.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04014235.hg.1 ENST00000226951.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014235.hg.1 ENST00000429087.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014235.hg.1 ENST00000442825.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014235.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014235.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014235.hg.1 TC04001036.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04014242.hg.1 ENST00000226951.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014242.hg.1 ENST00000442825.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014242.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014242.hg.1 TC04001036.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014244.hg.1 ENST00000226951.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014244.hg.1 ENST00000442825.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014244.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014244.hg.1 TC04001036.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04014236.hg.1 ENST00000226951.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014236.hg.1 ENST00000429087.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014236.hg.1 ENST00000442825.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014236.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014236.hg.1 NM_052964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014236.hg.1 TC04001036.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168932.hg.1 TC0400010050.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 ENST00000515667.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay TC0400010050.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168930.hg.1 TC0400010050.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 ENST00000442825.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 ENST00000507719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168931.hg.1 TC0400010050.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400168938.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168938.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168938.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168938.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168938.hg.1 TC0400010050.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400168940.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168940.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168940.hg.1 ENST00000226951.10 Ensembl
Affymetrix Human Clariom D Assay PSR0400168940.hg.1 NM_052964.3 Refseq
Affymetrix Human Clariom D Assay PSR0400168940.hg.1 TC0400010050.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 187395 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline