This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01076909_m1
See other CLNK GE Assays ›
Gene Symbol:
CLNK
Gene Name:
cytokine dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.4: 10489899 - 10734853 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01076909_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_052964.2 NP_443196.2 8 - 9 584 70
XM_011513775.2 XP_011512077.1 8 - 9 815 70
XM_017007684.1 XP_016863173.1 8 - 9 815 70
XM_017007685.1 XP_016863174.1 8 - 9 815 70
GenBank mRNA AK093001.1 - 8 - 9 586 70
BC029887.1 - 8 - 9 614 70
BC137553.1 - 8 - 9 513 9021553 70
BC137556.1 - 8 - 9 513 9021556 70

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLNK

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of signal transduction
intracellular signal transduction
SH3/SH2 adaptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline