This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01077025_m1
See other RPS6KA4 GE Assays ›
Gene Symbol:
RPS6KA4
Gene Name:
ribosomal protein S6 kinase A4
Gene Aliases:
MSK2, RSK-B, S6K-alpha-4
Chromosome Location:
Chr.11: 64359153 - 64372215 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01077025_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS6KA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318361.1 NP_001305290.1 2 - 3 168 109
GenBank mRNA AK298507.1 - 2 - 3 139 109

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK298507.1, NM_001318361.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine production
regulation of transcription, DNA-templated
protein phosphorylation
inflammatory response
histone phosphorylation
positive regulation of CREB transcription factor activity
positive regulation of histone phosphorylation
positive regulation of histone acetylation
intracellular signal transduction
histone H3-S10 phosphorylation
histone H3-S28 phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of NF-kappaB transcription factor activity
interleukin-1-mediated signaling pathway
magnesium ion binding
protein serine/threonine kinase activity
ribosomal protein S6 kinase activity
protein binding
ATP binding
histone kinase activity (H3-S10 specific)
mitogen-activated protein kinase p38 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline