This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01077248_m1
See other EHHADH GE Assays ›
Gene Symbol:
EHHADH
Gene Name:
enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
Gene Aliases:
ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE
Chromosome Location:
Chr.3: 185190624 - 185282886 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01077248_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EHHADH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001966.3 NP_001957.2 2 - 3 253 145
XM_006713525.1 XP_006713588.1 2 - 3 204 145
GenBank mRNA AK223460.1 - 2 - 3 217 145
AK291798.1 - 2 - 3 204 145
BC030982.1 - 2 - 3 204 145
BC038948.1 - 2 - 3 194 5533337 145
BC110460.2 - 2 - 3 184 40026238 145
L07077.1 - 2 - 3 184 145

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013408.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03002076.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_006713525.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EHHADH

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

internal protein amino acid acetylation
fatty acid beta-oxidation
cellular lipid metabolic process
3-hydroxyacyl-CoA dehydrogenase activity
dodecenoyl-CoA delta-isomerase activity
enoyl-CoA hydratase activity
receptor binding
protein binding
long-chain-enoyl-CoA hydratase activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000440662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000440662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000440662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000465178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000465178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000465178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 TC03002076.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 ENST00000231887.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 ENST00000456310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 ENST00000537544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 NM_001166415.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03036748.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay PSR0300199939.hg.1 TC0300013408.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000231887.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000440662.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000440662.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000440662.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000456310.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000465178.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000465178.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000465178.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000475987.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 ENST00000483104.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001166415.1 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq
Affymetrix Human Clariom D Assay TC0300013408.hg.1 NM_001966.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0