This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01078001_m1
See other NAA35 GE Assays ›
Gene Symbol:
NAA35
Gene Name:
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Gene Aliases:
EGAP, MAK10, MAK10P, bA379P1.1
Chromosome Location:
Chr.9: 85941142 - 86026355 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01078001_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024635.3 NP_078911.3 1 - 2 131 112
GenBank mRNA AK025266.1 - 1 - 2 127 112
AK225237.1 - 1 - 2 127 112
AK292239.1 - 1 - 2 131 112
DB023442.1 - 1 - 2 131 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900007775.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000391.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK025266.1,AK292239.1,NM_024635.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NAA35

Panther Classification:

Molecular Function -

acetyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
smooth muscle cell proliferation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09004179.hg.1 TC09000391.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09004179.hg.1 ENST00000361671.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004179.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004182.hg.1 TC09000391.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09004182.hg.1 ENST00000361671.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004182.hg.1 ENST00000376040.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004182.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004177.hg.1 TC09000391.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09004177.hg.1 ENST00000361671.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004177.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004178.hg.1 TC09000391.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09004178.hg.1 ENST00000361671.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004178.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152530.hg.1 TC0900007775.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900152529.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152529.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152529.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152529.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152529.hg.1 TC0900007775.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152531.hg.1 TC0900007775.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152535.hg.1 TC0900007775.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0