This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01078384_g1
See other APBB3 GE Assays ›
Gene Symbol:
APBB3
Gene Name:
amyloid beta precursor protein binding family B member 3
Gene Aliases:
FE65L2, SRA
Chromosome Location:
Chr.5: 140558268 - 140564604 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01078384_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APBB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133172.2 NP_573418.2 6 - 7 984 112
GenBank mRNA AF224708.1 - 6 - 7 645 112

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APBB3

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
beta-amyloid binding
protein binding
transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jrd88:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0