This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01078920_g1
See other NAA35 GE Assays ›
Gene Symbol:
NAA35
Gene Name:
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Gene Aliases:
EGAP, MAK10, MAK10P, bA379P1.1
Chromosome Location:
Chr.9: 85941142 - 86026355 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01078920_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321882.1 NP_001308811.1 1 - 2 346 82
GenBank mRNA AK056059.1 - 1 - 2 346 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK056059.1, NM_001321882.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
smooth muscle cell proliferation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0