| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000338963.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000435532.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000503468.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000506154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000508342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000508871.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_001284338.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_001284339.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_001284340.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_006154.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_198400.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NR_104302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
TC1500009508.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000338963.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000435532.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000503468.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000506154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000508342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
ENST00000508871.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_001284338.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_001284339.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_001284340.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_006154.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NM_198400.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500170186.hg.1 |
NR_104302.1 |
Refseq |