This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01081368_g1
See other DNMT3L GE Assays ›
Gene Symbol:
DNMT3L
Gene Name:
DNA methyltransferase 3 like
Gene Aliases:
-
Chromosome Location:
Chr.21: 44246339 - 44262216 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01081368_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNMT3L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013369.3 NP_037501.2 4 - 5 718 75
NM_175867.2 NP_787063.1 4 - 5 718 75
GenBank mRNA AF194032.1 - 4 - 5 718 75
BC002560.2 - 4 - 5 392 3138514 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100008356.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000517.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DNMT3L

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
regulation of gene expression by genetic imprinting
spermatogenesis
positive regulation of catalytic activity
negative regulation of transcription, DNA-templated
protein binding
enzyme activator activity
enzyme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21006217.hg.1 ENST00000270172.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21006217.hg.1 ENST00000418993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21006217.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21006217.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21006217.hg.1 TC21000517.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000436357.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay TC2100008356.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 TC2100008356.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 NM_175867.2 Refseq
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 ENST00000270172.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 ENST00000431166.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 ENST00000628202.2 Ensembl
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 NM_013369.3 Refseq
Affymetrix Human Clariom D Assay PSR2100156624.hg.1 NM_175867.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0