This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01081400_m1
See other TMPRSS15 GE Assays ›
Gene Symbol:
TMPRSS15
Gene Name:
transmembrane protease, serine 15
Gene Aliases:
ENTK, PRSS7
Chromosome Location:
Chr.21: 18269107 - 18403806 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01081400_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002772.2 NP_002763.2 12 - 13 1454 140
XM_011529654.1 XP_011527956.1 14 - 15 1728 140
XM_011529655.1 XP_011527957.1 15 - 16 1646 140
XM_011529656.2 XP_011527958.1 15 - 16 1653 140
XM_011529657.1 XP_011527959.1 13 - 14 1683 140
XM_011529658.1 XP_011527960.1 12 - 13 1647 140
XM_011529659.1 XP_011527961.1 13 - 14 1618 140
GenBank mRNA BC111749.1 - 12 - 13 1443 40008569 140
U09860.1 - 12 - 13 1463 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007676.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000305.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMPRSS15

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
receptor-mediated endocytosis
serine-type endopeptidase activity
scavenger receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21003650.hg.1 ENST00000284885.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21003650.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21003650.hg.1 TC21000305.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000474775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000474775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 ENST00000474775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000422787.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000474775.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000474775.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 ENST00000474775.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay TC2100007676.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152041.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100152041.hg.1 NM_002772.2 Refseq
Affymetrix Human Clariom D Assay PSR2100152041.hg.1 TC2100007676.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100152041.hg.1 ENST00000284885.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100152041.hg.1 NM_002772.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0